Terbium(III)kloridi, ultra dry ≥99.99% (REO, rare earth oxide basis)

Toimittaja: Thermo Fisher Scientific
Varoitus


Ampuled under argon

35798.03 35798.09
35798.03EA 245 EUR
35798.03 35798.09
Terbium(III)kloridi, ultra dry ≥99.99% (REO, rare earth oxide basis)
Terbium(III)kloridi

Purity: 99.99% CAS No.: 10042-88-3 Molecular Formula: TbCl3 Formula Weight: 265.29 Physical Form: Powder, Ampouled under argon Sensitivity: Hygroscopic Merck Reference: 14,9157 MDL No.: MFCD00011258

Kaava: Cl₃Tb
Sulamispiste: 588 °C
Tiheys: 4,35 g/cm³ (25 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011258
CAS-numero: 10042-88-3
EINECS: 233-132-4
Merck Index: 14,9157

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR