Molybdeeni standardiliuos 1000 mg/L Mo in 5% typpihapossa with hydrofluoric acid (max. 1%) (from Mo) Specpure® standard for AAS

Toimittaja: Thermo Fisher Scientific

35764.AE 35764.AP
35764.AEEA 104 EUR
35764.AE 35764.AP
Molybdeeni standardiliuos 1000 mg/L Mo in 5% typpihapossa with hydrofluoric acid (max. 1%) (from Mo) Specpure® standard for AAS
Standardiaineet ja vertailumateriaalit Cationic Standards Molybdenum Standards

Molecular Formula: Mo in 5% HNO₃/tr. HF
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00003465


Sertifikaatit: Traceable to NIST

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR