Volframi 1000 mg/L W in 5% typpihapossa with hydrofluoric acid (max. 1%) (from W) Specpure® standard for AAS

Toimittaja: Thermo Fisher Scientific

35760.AE 35760.AP
35760.AEEA 90.2 EUR
35760.AE 35760.AP
Volframi 1000 mg/L W in 5% typpihapossa with hydrofluoric acid (max. 1%) (from W) Specpure® standard for AAS
Standardiaineet ja vertailumateriaalit Cationic Standards Tungsten Standards

Molecular Formula: W in 5% HNO₃/tr. HF
Physical Form: Liquid
Odour: Acidic
UN No.: 3264
MDL No.: MFCD00011461


Sertifikaatit: Traceable to NIST

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR