Tallium(I)bromidi, dry_ultra 99,998 Metals basis

Toimittaja: Thermo Fisher Scientific
Vaara

Synonyms: Thallium bromide

ampuled under argon

35720.09 35720.18
35720.09EA 363 EUR
35720.09 35720.18
Tallium(I)bromidi, dry_ultra 99,998 Metals basis
Tallium(I)bromidi

Purity: 99.998% CAS No.: 7789-40-4 Molecular Formula: TlBr Formula Weight: 284.28 Physical Form: -10 Mesh Beads, Ampouled under argon Density: 7.5 Melting Point: ca 460° Merck Reference: 14,9258 UN No.: 1707 MDL No.: MFCD00011273

Kaava: BrTl
MW: 284.29 g/mol
Kiehumispiste: 815
Sulamispiste: ∼460 °C
Tiheys: 7.5
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011273
CAS-numero: 7789-40-4
EINECS: 232-163-0
UN: 1707
ADR: 6.1 II
Merck Index: 14,9258

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR