Kalsiumbromidi, ultra dry ≥99.978% (metals basis)

Toimittaja: Thermo Fisher Scientific
Varoitus


Ampuled under argon

35684.03 35684.06
35684.03EA 142 EUR
35684.03 35684.06
Kalsiumbromidi, ultra dry ≥99.978% (metals basis)
Kalsiumbromidi

Purity: 99.978% CAS No.: 7789-41-5 Molecular Formula: Br2Ca Formula Weight: 199.90 Physical Form: -10 Mesh Beads, Ampouled under argon Density: 3.353 Appearance: White to off-white Odour: Odourless Melting Point: 730° Boiling Point: 806-812° Merck Reference: 14,1654 MDL No.: MFCD00010902

Kaava: Br₂Ca
Kiehumispiste: 806…812 °C
Sulamispiste: 730 °C
Tiheys: 3,353
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00010902
CAS-numero: 7789-41-5
EINECS: 232-164-6
Merck Index: 14,1654

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR