Kadmiumjodidi, ultra dry ≥99.9985% (metals basis)

Toimittaja: Thermo Fisher Scientific
Vaara


Ampuled under argon

35683.03 35683.06 35683.14
35683.03EA 90.4 EUR
35683.03 35683.06 35683.14
Kadmiumjodidi, ultra dry ≥99.9985% (metals basis)
Kadmiumjodidi

Purity: 99.9985% CAS No.: 7790-80-9 Molecular Formula: CdI2 Formula Weight: 366.21 Physical Form: -10 Mesh Beads, Ampouled under argon Density: 5.67 Appearance: White to yellow Odour: Odourless Melting Point: 387-389° Boiling Point: 787° Merck Reference: 14,1621 UN No.: 2570 MDL No.: MFCD00010920

Kaava: CdI₂
Kiehumispiste: 787 °C
Sulamispiste: 387…389 °C
Tiheys: 5,67
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00010920
CAS-numero: 7790-80-9
EINECS: 232-223-6
UN: 2570
ADR: 6.1,III
Merck Index: 14,1621

Order Now

TEKNISET TIEDOT

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR