Rikkihappo standardised solution 0.5 N vesiliuos

Toimittaja: Thermo Fisher Scientific
Vaara

35653.K2 35653.M4
35653.K2EA 77.1 EUR
35653.K2 35653.M4
Rikkihappo standardised solution 0.5 N vesiliuos
Rikkihappo

Molecular Formula: H2SO4 Formula Weight: 98.08 Physical Form: Liquid Odour: Odourless UN No.: 2796 MDL No.: MFCD00064589

Kaava: H₂SO₄
Tiheys: 1,01 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00064589
CAS-numero: 7664-93-9
EINECS: 231-639-5
UN: 3264
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR