Typpihappo standardised solution 1.0 N

Toimittaja: Thermo Fisher Scientific
Vaara

Synonyms: Nitric acid solution

35624.AP 35624.K2
35624.APEA 93 EUR
35624.AP 35624.K2
Typpihappo standardised solution 1.0 N
Typpihappo

CAS No.: 7697-37-2 Molecular Formula: HNO3 Formula Weight: 63.01 Physical Form: Liquid Appearance: Colourless UN No.: 3264 MDL No.: MFCD00011349

Kaava: HNO₃
MW: 63.01 g/mol
Kiehumispiste: 87
Sulamispiste: -42
Tiheys: 1.035 g/cm³ (25 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011349
CAS-numero: 7697-37-2
EINECS: 231-714-2
UN: 3264
ADR: 8 III
Merck Index: 13,06610

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR