Hopeanitraatti standardised solution 1.0 N vesiliuos

Toimittaja: Thermo Fisher Scientific
Vaara

35597.AP 35597.K2
35597.APEA 1620 EUR
35597.AP 35597.K2
Hopeanitraatti standardised solution 1.0 N vesiliuos
Hopeanitraatti

CAS No.: 7761-88-8 Molecular Formula: AgNO3 Formula Weight: 169.87 Physical Form: Liquid Sensitivity: Light Sensitive Appearance: Colourless UN No.: 3264 MDL No.: MFCD00003414

Kaava: AgNO₃
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00003414
CAS-numero: 7761-88-8
EINECS: 231-853-9
UN: 3264
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR