Kaliumjodaatti standardised solution 0.1 N vesiliuos

Toimittaja: Thermo Fisher Scientific

35586.K2 35586.K7
35586.K2EA 132 EUR
35586.K2 35586.K7
Kaliumjodaatti standardised solution 0.1 N vesiliuos
Kaliumjodaatti

CAS No.: 7758-05-6 Molecular Formula: KIO3 Formula Weight: 214.00 Physical Form: Liquid Appearance: Colourless Odour: Odourless MDL No.: MFCD00011406

Kaava: KlO₃
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011406
CAS-numero: 7758-05-6
EINECS: 231-831-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR