Asetonitriili ≥99.8% (by GC), CHROMASOLV™ for liquid chromatography, Riedel-de Haën™

Toimittaja: Honeywell Chemicals
Vaara

34881-18L 34881-45L 34881-7L 34881-1L 34881-2.5L 34881-4X2.5L 34881-30L-RC
34881-18LEA 2970 EUR
34881-18L 34881-45L 34881-7L 34881-1L 34881-2.5L 34881-4X2.5L 34881-30L-RC
Asetonitriili ≥99.8% (by GC), CHROMASOLV™ for liquid chromatography, Riedel-de Haën™
Asetonitriili

  • For GC headspace applications
  • Meets USP, Ph. Eur. and ICH requirements
  • GC headspace application tested
  • Packed under inert gas for longer shelf life

Kaava: H₃CCN
Kiehumispiste: 82 °C (1,013 hPa)
Sulamispiste: –46 °C
Tiheys: 0,7857 g/cm³ (20 °C)
Leimahduspiste: 6 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00001878
CAS-numero: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3,II
REACH: 01-2119471307-38
Merck Index: 13,00071
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Refractive Index n20/D 1.344 (lit.)
Assay (GC) Min. 99.8 %
Non-volatile matter Max. 0.0005 %
Water (Karl Fischer) Max. 0.1 %
Free acid (as CH₃COOH) Max. 0.001 %
Transmittance at 200 nm Min. 50 %
Transmittance at 220 nm Min. 90 %
Transmittance from 240 nm Min. 99 %
Vapor Pressure 72.8 mmHg ( 20 °C)
Explosion Limit 16 %
Autoignition Temperature 973 °F


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR