Kloroformi ≥99.8% stabilised, CHROMASOLV™ for HPLC, Riedel-de Haën™

Toimittaja: Honeywell Chemicals
Vaara


Stabilization: Stabilised with amylene

34854-1L 34854-2.5L 34854-4X2.5L 34854-6X1L
34854-1LEA 125 EUR
34854-1L 34854-2.5L 34854-4X2.5L 34854-6X1L
Kloroformi ≥99.8% stabilised, CHROMASOLV™ for HPLC, Riedel-de Haën™
Kloroformi

Chloroform or Trichloromethane, is an organic compound with formula CHCl3. It is a colorless, sweet-smelling, dense liquid. Suitable for HPLC and spectrophotometry.


  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency

Kaava: CHCl₃
Kiehumispiste: 61 °C (1,013 hPa)
Tiheys: 1,485 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00000826
CAS-numero: 67-66-3
EINECS: 200-663-8
UN: 1888
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cc00da8cefd4e04c0ea1d7c5801308cd73eeef9&vid=c7d46a2ee73651eae0f3654c272e820106b2f04f&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Assay (GC) Min. 99.80 %
Non-volatile matter Max. 0.0005 %
Water (Karl Fischer) Max. 0.01 %
Free acid (as HCl) Max. 0.001 %
Absorbance at 250 nm Max. 1.00
Absorbance at 260 nm Max. 0.30
Absorbance 280 nm Max. 0.005
IR spectrum complying
Vapor Density 4.1 (vs air)
Refractive Index n20/D 1.445 (lit.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR