Reagents for water determination by the Coulometric Titration Karl Fischer method, Hydranal™
Toimittaja: Honeywell Chemicals
34821-50ML 34836-500ML 34807-500ML 34810-500ML 34820-500ML 34843-500ML 34739-500ML 34829-1L 34840-50ML 34726-500ML 34836-1L 34726-6X500ML 34739-6X500ML 34807-6X500ML 34810-6X500ML 34820-6X500ML 34829-6X1L 34836-6X1L 34836-6X500ML 34843-6X500ML 34821-6X50ML 34840-6X50ML
34821-50MLEA 304.3 EUR
34821-50ML 34836-500ML 34807-500ML 34810-500ML 34820-500ML 34843-500ML 34739-500ML 34829-1L 34840-50ML 34726-500ML 34836-1L 34726-6X500ML 34739-6X500ML 34807-6X500ML 34810-6X500ML 34820-6X500ML 34829-6X1L 34836-6X1L 34836-6X500ML 34843-6X500ML 34821-6X50ML 34840-6X50ML
Reagents for water determination by the Coulometric Titration Karl Fischer method, Hydranal™
Karl Fischer-titrantit ja -reagenssit
Coulometric Karl Fischer titrations normally require two reagent solutions: an anolyte (the solution in the anodic compartment) and a catholyte (the solution in the cathodic compartment).
  • Easy to use
  • High accuracy for trace amounts of water
  • Stable conditions of the titration vessel
  • Broad product range available
  • Long shelf life (up to five years)