Natriumasetaatti, vedetön ≥99.0%, BAKER ANALYZED® ACS, J.T.Baker®

Toimittaja: Avantor

3470-01EA 206.38 EUR
3470-01 3470-07 3470-R 3470-05
Natriumasetaatti, vedetön ≥99.0%, BAKER ANALYZED® ACS, J.T.Baker®
Natriumasetaatti

  • Lot analysis on label

Kaava: H₃CCOONa
Kiehumispiste: 881,4 °C (1013 hPa)
Sulamispiste: 324 °C
Tiheys: 1,53 g/cm³
Leimahduspiste: >250 °C (sulj. astia)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00012459
CAS-numero: 127-09-3
EINECS: 204-823-8
REACH: 01-2119485123-42
Merck Index: 12,08711

Order Now

Testitulosten erittely

Meets ACS Reagent Chemical Requirements ≥99.0%
For Laboratory,Research,or Manufacturing Use ≤0.01%
ACS - Assay (CH₃COONa) ≥99.0 %
ACS - Insoluble Matter ≤0.01 %
ACS - Loss on Drying at 120°C ≤1.0 %
ACS - pH of 5% Solution at 25°C 7.0 - 9.2
ACS - Chloride (Cl) ≤0.002 %
ACS - Phosphate (PO₄) ≤0.001 %
ACS - Sulfate (SO₄) ≤0.003 %
ACS - Calcium (Ca) ≤0.005 %
ACS - Heavy Metals (as Pb) ≤0.001 %
ACS - Iron (Fe) ≤0.001 %
ACS - Magnesium (Mg) ≤0.002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR