Hopeasulfaatti 10 g/l in sulphuric acid mittaliuos, for COD determination according to DIN 38409-41, Fluka™

Toimittaja: Honeywell Chemicals
Vaara

34629-1L 34629-2.5L 34629-6X1L 34629-4X2.5L
34629-1LEA 116 EUR
34629-1L 34629-2.5L 34629-6X1L 34629-4X2.5L
Hopeasulfaatti 10 g/l in sulphuric acid mittaliuos, for COD determination according to DIN 38409-41, Fluka™
Hopeasulfaatti

  • Solutions are prepared with raw materials which meet or exceed requirements of the American Chemical Society
  • Where no ACS specifications exist, chemicals of highest purity available are used
  • All aqueous solutions are prepared using high purity, deionized, water which meets requirements for ASTM type 1 Reagent Grade Water
  • Exacting production and quality control records are maintained providing traceability of raw materials
  • All solutions are guaranteed to meet label specifications
  • All solution labels meet or exceed all ISO, DOT and OSHA regulations

Kaava: Ag₂SO₄
Kiehumispiste: 338 °C (1,013 hPa) (main component)
Tiheys: 1,85 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00003407
CAS-numero: 10294-26-5
EINECS: 233-653-7
UN: 3264
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd7afaa8e9ae24bf040d40aed0c93763ce897080&vid=ae7b50634b53507047974c2157e5e38c2eb90a20&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Assay of H₂SO₄ 94 - 96 %
Assay of Ag₂SO₄ 0.54 - 0.56 %
Density (20°C) 1.83 - 1.85 g/ml


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR