Ferroin (1,10-phenanthrolineferrous sulphate complex) solution Reag. Ph. Eur. redox indicator E0 (H2SO4 1 mol/l) = +1,06 volt, Fluka™

Toimittaja: Honeywell Chemicals

34531-100ML 34531-500ML
34531-100MLEA 77.5 EUR
34531-100ML 34531-500ML
Ferroin (1,10-phenanthrolineferrous sulphate complex) solution Reag. Ph. Eur. redox indicator E0 (H2SO4 1 mol/l) = +1,06 volt, Fluka™
Ferroin (1,10-phenanthrolineferrous sulphate complex)

E0 in sulfuric acid 1 mol/l = +1,06 volt

Kaava: C₃₆H₂₄FeN₆SO₄
Kiehumispiste: 100 °C (1,013 hPa)
Tiheys: 1,00 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00036428
CAS-numero: 14634-91-4

Order Now

Testitulosten erittely

Change complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR