Metanoli ≥99.9% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™

Toimittaja: Honeywell Chemicals
Vaara

34485-7L 34485-1L 34485-2.5L 34485-4X2.5L
34485-7LEA 328 EUR
34485-7L 34485-1L 34485-2.5L 34485-4X2.5L
Metanoli ≥99.9% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Metanoli

  • For gas chromatography (GC)
  • Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
  • Non-volatile matter and water content are specified for each solvent

Kaava: CH₃OH
Kiehumispiste: 64…65 °C (1,013 hPa)
Sulamispiste: –98 °C
Tiheys: 0,79 g/cm³ (20 °C)
Leimahduspiste: 11 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00004595
CAS-numero: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3,II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Assay (GC) Min. 99.9 %
Non-volatile matter Max. 0.0005 %
Water (Karl Fischer) Max. 0.05 %
Suitability for residue analysis Max. 5 ng/l
Vapor Pressure 97.68 mmHg ( 20 °C)
Refractive Index n20/D 1.329 (lit.)
Explosion Limit 36 %
Autoignition Temperature 725 °F


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR