Bromitymolisininen 0,04% etanolissa, TECHNICAL

Toimittaja: VWR Chemicals
Vaara

34303.234EA 81.3 EUR
34303.234
Bromitymolisininen 0,04% etanolissa, TECHNICAL
Bromitymolisininen

  • Transition range: pH 6,0-7,6

Kaava: C₂₇H₂₈Br₂O₅S
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00005872
CAS-numero: 76-59-5
EINECS: 200-971-2
UN: 1170
ADR: 3,II

Order Now

Testitulosten erittely

Identification Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR