Sinkkisulfaatti-7 H2O 99.0-103.0% ACS

Toimittaja: Thermo Fisher Scientific
Vaara

33399.22 33399.36 33399.A4
33399.22EA 54.1 EUR
33399.22 33399.36 33399.A4
Sinkkisulfaatti-7 H2O 99.0-103.0% ACS
Sinkkisulfaatti-7 H2O

Purity: 99.0-103.0% CAS No.: 7446-20-0 Molecular Formula: O4SZn.7H2O Formula Weight: 287.54 (161.43anhy) Physical Form: Lump Density: 1.97 Appearance: Colourless to white pH: 4.4-6 Melting Point: 100° Solubility: Soluble in water, glycerol Merck Reference: 14,10159 UN No.: 3077 MDL No.: MFCD00149894

Kaava: ZnSO₄·7H₂O
Sulamispiste: 100 °C
Tiheys: 1,97
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00149894
CAS-numero: 7446-20-0
EINECS: 231-793-3
UN: 3077
ADR: 9,III
Merck Index: 14,10159

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR