Natriumhypokloriitti (11 - 15% Cl₂)

Toimittaja: Thermo Fisher Scientific
Vaara

33369.36 33369.A3
33369.36EA 102 EUR
33369.36 33369.A3
Natriumhypokloriitti (11 - 15% Cl₂)
Natriumhypokloriitti

CAS No.: 7681-52-9 Molecular Formula: ClNaO Formula Weight: 74.44 Storage Temperature: 2°C to 8°C Physical Form: Liquid Density: 1.2 Appearance: Clear yellow or green Merck Reference: 14,8628 UN No.: 1791 MDL No.: MFCD00011120

Kaava: ClNaO
Kiehumispiste: 111 °C (1013 hPa)
Sulamispiste: –30…–20 °C
Tiheys: 1,2
Storage Temperature: Jääkaappi
MDL numero: MFCD00011120
CAS-numero: 7681-52-9
EINECS: 231-668-3
UN: 1791
ADR: 8,III
Merck Index: 14,8628

Order Now

Testitulosten erittely

Appearance (Colour) Pale yellow to green
Assay (unspecified) 10,5 - 15,5% available chlorine
Form Liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR