Rauta(III)sulfaatti-hydraatti, Reagent Grade

Toimittaja: Thermo Fisher Scientific
Varoitus

33316.A7 33316.30 33316.A1
33316.A7EA 907 EUR
33316.A7 33316.30 33316.A1
Rauta(III)sulfaatti-hydraatti, Reagent Grade
Rauta(III)sulfaatti-hydraatti

CAS No.: 15244-10-7 Molecular Formula: Fe2(SO4)3.xH2O Formula Weight: 399.87(anhy) Physical Form: Granular/Powder Density: 2.0-2.1 Sensitivity: Hygroscopic Appearance: Grayish white Solubility: Soluble in water MDL No.: MFCD00011007

Kaava: Fe₂(SO₄)₃·xH₂O
Tiheys: 2,0…2.1
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00149714
CAS-numero: 15244-10-7
EINECS: 233-072-9
Merck Index: 13,04065

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR