Rauta (III) nitraatti-9 H2O 98.0-101.0% ACS

Toimittaja: Thermo Fisher Scientific
Vaara

33315.A3 33315.36 33315.F7 33315.PG
33315.A3EA 392 EUR
33315.A3 33315.36 33315.F7 33315.A7
Rauta (III) nitraatti-9 H2O 98.0-101.0% ACS
Rauta (III) nitraatti-9 H2O

Purity: 98.0-101.0% CAS No.: 7782-61-8 Molecular Formula: FeN3O9.9H2O Formula Weight: 403.99 (241.86anhy) Physical Form: Crystalline Density: 1.684 Appearance: Pale purple Odour: Acidic Melting Point: 47° Boiling Point: 125° dec. Solubility: Freely soluble in water, alcohol, acetone. Slightly soluble in cold and concentrated HNO3 Merck Reference: 14,4027 UN No.: 1466 MDL No.: MFCD00149708

Kaava: Fe(NO₃)₃·9H₂O
Kiehumispiste: 125 °C (haj. lämpötila)
Sulamispiste: 47 °C
Tiheys: 1,684
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00149708
CAS-numero: 7782-61-8
EINECS: 233-899-5
UN: 1466
ADR: 5.1,III
Merck Index: 14,4027

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR