Chromium (III) sulphate hydrate, reagent

Toimittaja: Thermo Fisher Scientific
Varoitus

33303.22 33303.36
33303.22EA 209 EUR
33303.22 33303.36
Chromium (III) sulphate hydrate, reagent
Chromium (III) sulphate hydrate

CAS No.: 15244-38-9 Molecular Formula: Cr2(SO4)3.xH2O Formula Weight: 392.17(anhy) Physical Form: Crystalline Sensitivity: Hygroscopic Appearance: Green to violet Odour: Odourless MDL No.: MFCD00016039

Kaava: Cr₂O₁₂S₃·H₂O
MW: 392.17 g/mol (anhy)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00016039
CAS-numero: 15244-38-9
EINECS: 233-253-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR