Etikkahappo ≥99.8%, Puriss. p.a. Reag. Ph. Eur., ACS, ISO, Fluka™

Toimittaja: Honeywell Chemicals
Vaara

Synonyms: Vinegar acid, Methanecarboxylic acid, Ethanoic acid

33209-1L 33209-1L-GL 33209-2.5L 33209-2.5L-GL 33209-30KG 33209-4X2.5L 33209-4X2.5LGL 33209-60KG 33209-6X1L 33209-5L 33209-200L
33209-1LEA 63.56 EUR
33209-1L 33209-1L-GL 33209-2.5L 33209-2.5L-GL 33209-30KG 33209-4X2.5L 33209-4X2.5LGL 33209-60KG 33209-6X1L 33209-5L 33209-200L
Etikkahappo ≥99.8%, Puriss. p.a. Reag. Ph. Eur., ACS, ISO, Fluka™
Etikkahappo
Kaava: H₃CCOOH
Kiehumispiste: 118 °C (1,013 hPa)
Sulamispiste: 16 °C
Tiheys: 1,05 g/cm³ (20 °C)
Leimahduspiste: 40 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00036152
CAS-numero: 64-19-7
EINECS: 200-580-7
UN: 2789
ADR: 8,II
REACH: 01-2119475328-30
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Assay 99.8 - 100.5 %
Assay (GC) Min. 99.8 %
Density (D 20/20) 1.052 - 1.053
Boiling range 117 - 119 °C
Congealing - Freezing Point Min. 16.3 °C
Non-volatile matter Max. 0.0005 %
Water (Karl Fischer) Max. 0.4 %
Silver (Ag) Max. 0.000001 %
Aluminium (Al) Max. 0.000005 %
Arsenic (As) Max. 0.000001 %
Barium (Ba) Max. 0.000001 %
Beryllium (Be) Max. 0.000001 %
Bismuth (Bi) Max. 0.00001 %
Calcium (Ca) Max. 0.00002 %
Cadmium (Cd) Max. 0.000002 %
Cobalt (Co) Max. 0.000001 %
Chromium (Cr) Max. 0.000005 %
Copper (Cu) Max. 0.000001 %
Iron (Fe) Max. 0.00002 %
Germanium (Ge) Max. 0.000005 %
Potassium (K) Max. 0.00001 %
Lithium (Li) Max. 0.000001 %
Magnesium (Mg) Max. 0.00001 %
Manganese (Mn) Max. 0.000001 %
Molybdenum (Mo) Max. 0.000002 %
Sodium (Na) Max. 0.00005 %
Nickel (Ni) Max. 0.000005 %
Lead (Pb) Max. 0.000002 %
Strontium (Sr) Max. 0.000001 %
Titanium (Ti) Max. 0.000001 %
Thallium (Tl) Max. 0.000005 %
Vanadium (V) Max. 0.000001 %
Zinc (Zn) Max. 0.000005 %
Zirconium (Zr) Max. 0.00001 %
Heavy metals (as Pb) Max. 0.00005 %
Chloride (Cl) Max. 0.00005 %
Phosphate (PO₄) Max. 0.00005 %
Sulfate (SO₄) Max. 0.00005 %
Acetaldehyde Max. 0.0002 %
Acetic aNHydride (GC) Max. 0.01 %
Mixable with H₂O complying
Titrable Base Max. 0.0004 meq/g
APHA Max. 10
Appearance of the substance complying
K₂Cr₂O₇ reducing matter complying
KMnO₄ reducing substances complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR