Sampling scoop, long handle, disposable Bio
Toimittaja: BURKLE
5379-1024 5379-0024 5379-1025 5379-0025
331-0354EA 61.7 EUR
331-0354 331-0355 331-0356 331-0357
Sampling scoop, long handle, disposable Bio
Kauhat
The sampling scoop can be used to take samples of powders, granulates and pastes. The long handle makes it possible to sample larger containers even with little content or to reach through narrow openings. With the straight bottom of the scoop, the medium can be scooped up directly from the bottom of the container.
  • Product made from renewable raw materials
  • Bio-PE, white
  • Individually packaged and optionally sterilised
  • Handle length 300 mm
  • For single use
  • Clean room manufactured (clean room class 7)
  • Complies with EU food requirements and FDA regulations