Sampling spoons, long handle, disposable, LaboPlast®/SteriPlast®
Toimittaja: BURKLE
5378-5002 5378-5001 5378-5006 5378-5005
331-0323EA 40 EUR
331-0323 331-0324 331-0325 331-0326
Sampling spoons, long handle, disposable, LaboPlast®/SteriPlast®
Lusikat
This sampling spoon with long handle can be used to remove powders, granulates, pastes or liquids.
  • PS, white
  • Individually packed and optionally sterilised
  • Handle length: 300 mm
  • For single use
  • Clean room manufactured (clean room Class 7)
The long handle makes it easy to reach the bottoms of larger containers as well as sampling of drums, jerricans and bottles with small opening diameters. Thanks to the indentations on the handle, the disposable spoon rests comfortably and securely in the hand. The laboratory spoon is suitable for single-use sensitiveMore Product Information