VWR® Standard Line, Pullo, kapea suuaukko, kierrekorkilla
Toimittaja: VWR Collection
331-0094EA 147 EUR
331-0094 331-0095 331-0096 331-0098 331-0099 331-0295 331-0296 331-0297 331-0317 331-0318 331-0319
VWR® Standard Line, Pullo, kapea suuaukko, kierrekorkilla
Pullot Muovipullot
PET, with or without PP screw caps.
  • 28 mm neck for all bottles
  • Green colour protects light-sensitive material
  • Manufactured from food grade PET
For basic sample collecting. Non sterile, lightweight and round.
Varoitus: The product(s) on this page are outside the scope of the EU Directive 2019/904.