Crucibles, Tammann, aluminum oxide
Toimittaja: Thermo Fisher Scientific
33052.KF 33053.KF 33055.KF 33056.KF 33058.KF
33052.KFEA 105 EUR
33052.KF 33053.KF 33055.KF 33056.KF 33058.KF
Crucibles, Tammann, aluminum oxide
Upokkaat
These crucibles according to Tammann are manufactured from aluminium oxide. They display superior characteristics both at elevated temperature and at high frequencies, and are resistant to corrosive liquids and gases. Oxide ceramics are also inert to oxidation and are not subject to radiation damage.