Human processed serum (HPS), Vitamin D depleted

Toimittaja: SEQENS
33000D-100ML 33000D-500ML 33000D-1L
33000D-100MLEA 206 EUR
33000D-100ML 33000D-500ML 33000D-1L
Human processed serum (HPS), Vitamin D depleted
Seerumit
SEQENS IVD manufactures Vitamin D Depleted Human Processed Serum.

  • Dry extract: 65 to 85 g/L
  • Total proteins: 55 to 75 g/L
  • Vitamin 25-OH D3: <4 ng/ml
  • Filtered through a 0,2 µm membrane
  • Shelf life: 2 years

For this product, source material has been tested and found negative or non-reactive (FDA approved and/or CE marked test methods) for anti-HIV 1+2 and anti-HCV antibodies, HBs antigen, HIV 1 and HCV RNA and Syphilitic serology.

Complete donor traceability on file.

Varoitus: Only for research or manufacture of in vitro diagnostic reagents. NotMore Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR