Rose Bengal 95% (dye content) ≥96% (by HPLC), Sigma-Aldrich®

Toimittaja: SIGMA ALDRICH MICROSCOPY

330000-1G 330000-5G
330000-1GEA 95.8 EUR
330000-1G 330000-5G
Rose Bengal 95% (dye content) ≥96% (by HPLC), Sigma-Aldrich®
Rose Bengal

Rose Bengal has been used to evaluate acrosome integrity. It has also been used to initiate photothrombotic occlusion in blood.

Kaava: C₂₀H₂Cl₄I₄O₅Na₂
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00151169
CAS-numero: 632-69-9
EINECS: 211-183-3

Order Now

Testitulosten erittely

Appearance (Colour) Red to Brown
Appearance (Form): Powder and/or Chunks Conforms
Infrared spectrum Conforms to Structure
Wave Length, c = 0.004 g/L H2O + 1 ml 1% Na2CO3 546 - 550 nm
Extinction Coefficient ≥32000
Wave Length 262 - 266 nm
Purity (HPLC) Approximately ≥95.0%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR