Crucibles, cylindrical, flat base, aluminum oxide (Al-23)
Toimittaja: Thermo Fisher Scientific
32950.KF 32951.KF 32952.KF 32953.KF 32954.KF 32955.KF 32957.KF
32950.KFEA 127 EUR
32950.KF 32951.KF 32952.KF 32953.KF 32954.KF 32955.KF 32957.KF
Crucibles, cylindrical, flat base, aluminum oxide (Al-23)
Upokkaat
These crucibles are manufactured from aluminium oxide. They display superior characteristics both at elevated temperature and at high frequencies, and are resistant to corrosive liquids and gases. Oxide ceramics are also inert to oxidation and are not subject to radiation damage.
  • 1950 °C (max service temp in air)