Rods, ceramic, round, solid, AL23
Toimittaja: Thermo Fisher Scientific
32921.DM 32931.DR 32931.DT 32931.DW 32931.EH 32933.DR
32921.DMEA 45.6 EUR
32921.DM 32931.DR 32931.DT 32931.DW 32931.EH 32933.DR
Rods, ceramic, round, solid, AL23
Sauvat
Pure Al₂O₃ Aluminium oxide, AL23.
Dense, excellent thermal and electrical resistance properties, corrosion resistant, permeable to microwaves.