NBQX 98+%

Toimittaja: Thermo Fisher Scientific
Varoitus

329260250 329261000
329260250.EA 510 EUR
329260250. 329261000.
NBQX 98+%
NBQX
Kaava: C₁₂H₈N₄O₆S
Kiehumispiste: 613°C
Storage Temperature: Jääkaappi
MDL numero: MFCD12910445
CAS-numero: 118876-58-7

Order Now

Testitulosten erittely

Appearance almost-white to yellow powder
Infrared spectrum Conforms
HPLC ≥98 %
Water ≤5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR