Fish spine beads, AL23
Toimittaja: Thermo Fisher Scientific
32905.KX 32906.KX 32907.KX 32908.KX
32905.KXEA 344 EUR
32905.KX 32906.KX 32907.KX 32908.KX
Fish spine beads, AL23
Helmet ja partikkelit Ceramic Beads
Pure Al₂O₃ Aluminium oxide, AL 23.
Dense, excellent thermal and electrical resistance properties, corrosion resistant, permeable to microwaves.