Manganese(II) iodide, anhydrous 98%

Toimittaja: Thermo Fisher Scientific
Varoitus

Synonyms: Manganous iodide

32808.09 32808.18
32808.09EA 221 EUR
32808.09 32808.18
Manganese(II) iodide, anhydrous 98%
Manganese(II) iodide

Purity: 98% CAS No.: 7790-33-2 Molecular Formula: I2Mn Formula Weight: 308.75 Physical Form: Powder Density: 5.0 Appearance: White to pink to red Odour: Odourless Melting Point: 638° Solubility: Soluble in water with gradual decomposition. Soluble in alcohol Merck Reference: 14,5732 MDL No.: MFCD00016225

Kaava: I₂Mn
MW: 308.75 g/mol
Sulamispiste: 638
Tiheys: 5.0
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00016225
CAS-numero: 7790-33-2
EINECS: 232-201-6
Merck Index: 14,5732

Order Now

TEKNISET TIEDOT

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR