6-O-Palmitoyl-L-ascorbic acid ≥99%

Toimittaja: Thermo Fisher Scientific

32791.14 32791.22 32791.36
32791.14EA 99 EUR
32791.14 32791.22 32791.36
6-O-Palmitoyl-L-ascorbic acid ≥99%
6-O-Palmitoyl-L-ascorbic acid

Purity: 99% CAS No.: 137-66-6 Molecular Formula: C22H38O7 Formula Weight: 414.52 Physical Form: Crystalline Odour: Pleasant MDL No.: MFCD00005377

Kaava: C₂₂H₃₈O₇
Kiehumispiste: ∼250 °C
Sulamispiste: 113…114 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00005377
CAS-numero: 137-66-6
EINECS: 205-305-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR