Organic reference standard, Patulin, Supelco®
Toimittaja: Merck
32759-5MG
32759-5MGEA 720 EUR
32759-5MG
Organic reference standard, Patulin, Supelco®
Standardiaineet ja vertailumateriaalit Organic Standards
Patulin (PAT) is a mycotoxin of the polyketide lactone type, usually produced by species of the Penicillium, Aspergillus and Byssochlamys genera.
Fruit-based products by QuEChERS (Quick, Easy, Cheap, Effective, Rugged and Safe) method and solid phase extraction (SPE) followed by analysis using ultra-high performance liquid chromatography combined with electrospray ionisation tandem mass spectrometry (UHPLC-ESI-MS/MS).
PAT may be used as a reference standard for the determination of patulin in model food matrices by 'dilute and shoot' LC-MS/MS, dairy products by QuEChERS procedure and UHPLC-ESI quadrupole orbitrap (Q-Orbitrap) mass spectrometry and apple juice samples by single drop liquid-liquid-liquid microextraction (SD-LLLME) followed by LC-MS/MS analysis.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5400c3ca4c9ff4d31da4f2acb5ad32f1d526a879&vid=9e99947409d8733bd56384b98c82901ec5cec0e7&prid=web&clid=SAPDAM