Bromitymolisininen sultone form ACS, Reag. Ph. Eur. indicator, Fluka™

Toimittaja: Honeywell Chemicals

32714-1KG 32714-5G 32714-25G
32714-1KGEA 2610 EUR
32714-1KG 32714-5G 32714-25G
Bromitymolisininen sultone form ACS, Reag. Ph. Eur. indicator, Fluka™
Bromitymolisininen
Kaava: C₂₇H₂₈Br₂O₅S
Sulamispiste: 204…202 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00005872
CAS-numero: 76-59-5
EINECS: 200-971-2
Merck Index: 13,01430

Order Now

Testitulosten erittely

Suitability as indicator complying
Test of sensitivity complying
Clearness of the solution complying
Change Range (pH6,0-7,4) complying
Curve course (UV) complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR