Organic reference standard, Melamine-13C3, analytical standard, VETRANAL™, Supelco®
Toimittaja: Merck
32666-10MG
32666-10MGEA 718 EUR
32666-10MG
Organic reference standard, Melamine-13C3, analytical standard, VETRANAL™, Supelco®
Standardiaineet ja vertailumateriaalit Organic Standards
Melamine-¹³C₃ may be used as an analytical standard for the determination of melamine in milk powder samples by liquid chromatography-electrospray ionisation-isotope dilution mass spectrometry (LC-ESI-IDMS).
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5902b7f454b6d326efd03fc08f0a737f4c198622&vid=5902b7f454b6d326efd03fc08f0a737f4c198622&prid=web&clid=SAPDAM