Bromifenolisininen ACS

Toimittaja: Thermo Fisher Scientific

Synonyms: Bromphenol Blue Sultone Form, BPB, 3,3',5,5'-Tetrabromophenolsulphonphthalein, Tetrabromophenol-sulphonephthalein

32641.03 32641.06 32641.14
32641.03EA 48.2 EUR
32641.03 32641.06 32641.14
Bromifenolisininen ACS
Bromifenolisininen

CAS No.: 115-39-9 Molecular Formula: C19H10Br4O5S Formula Weight: 669.98 Physical Form: Crystalline Odour: Odourless Melting Point: 270° dec. Beilstein Reference: 61698 Merck Reference: 14,1444 MDL No.: MFCD00005875

Kaava: C₁₉H₁₀Br₄O₅S
MW: 669.97 g/mol
Sulamispiste: 270 °C (haj. lämpötila)
Tiheys: 1 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00005875
CAS-numero: 115-39-9
EINECS: 204-086-2
Merck Index: 14,1444

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR