n-Heksaani ≥95% spektrofotometriaan

Toimittaja: Thermo Fisher Scientific
Vaara

32454.K2 32454.K7 32454.M1
32454.K2EA 226 EUR
32454.K2 32454.K7 32454.M1
n-Heksaani ≥95% spektrofotometriaan
n-Heksaani

Purity: 95+% CAS No.: 110-54-3 Molecular Formula: CH3(CH2)4CH3 Formula Weight: 86.18 Flash Point: -23°(-9°F) Physical Form: Liquid Density: 0.659 Refractive Index: 1.3750 Odour: Weak, characteristic Melting Point: -95° Boiling Point: 69° Beilstein Reference: 1730733 Merck Reference: 14,4694 UN No.: 1208 MDL No.: MFCD00009520

Kaava: H₃C(CH₂)₄CH₃
Kiehumispiste: 69 °C
Sulamispiste: –95 °C
Tiheys: 0,659
Leimahduspiste: –23 °C (–9 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00009520
CAS-numero: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3,II
Merck Index: 14,4694
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR