n-Pentaani 99+% spektrofotometriaan

Toimittaja: Thermo Fisher Scientific
Vaara

32449.K2 32449.K7 32449.M1 32449.M6
32449.K2EA 216 EUR
32449.K2 32449.K7 32449.M1 32449.M6
n-Pentaani 99+% spektrofotometriaan
n-Pentaani

Purity: 99+% min. CAS No.: 109-66-0 Molecular Formula: CH3(CH2)3CH3 Formula Weight: 72.15 Flash Point: -49°(-57°F) Physical Form: Liquid Density: 0.626 Refractive Index: 1.3580 Appearance: Colourless Melting Point: -130° Boiling Point: 36° Merck Reference: 14,7116 UN No.: 1265 MDL No.: MFCD00009498

Kaava: H₃C(CH₂)₃CH₃
Kiehumispiste: 36 °C
Sulamispiste: –130 °C
Tiheys: 0,626
Leimahduspiste: –49 °C (–57 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00009498
CAS-numero: 109-66-0
EINECS: 203-692-4
UN: 1265
ADR: 3,II
REACH: 01-2119459286-30
Merck Index: 14,7116
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ba2c9c81a4b1f44769627a00ba49b6cc8e63132&vid=f6831eb15ab1602d8c752468dc3ee31cc4ee61c0&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR