n-Heptaani 99+% spektrofotometriaan

Toimittaja: Thermo Fisher Scientific
Vaara

32441.K2 32441.M1
32441.K2EA 212 EUR
32441.K2 32441.M1
n-Heptaani 99+% spektrofotometriaan
n-Heptaani

Purity: 99+% CAS No.: 142-82-5 Molecular Formula: CH3(CH2)5CH3 Formula Weight: 100.21 Flash Point: -4°(24°F) Physical Form: Liquid Density: 0.684 Refractive Index: 1.3877 Odour: Petroleum-like Melting Point: -91° Boiling Point: 97-99° Beilstein Reference: 1730763 Merck Reference: 14,4659 UN No.: 1206 MDL No.: MFCD00009544

Kaava: H₃C(CH₂)₅CH₃
Kiehumispiste: 97…99 °C
Sulamispiste: –91 °C
Tiheys: 0,684
Leimahduspiste: –4 °C (24 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00009544
CAS-numero: 142-82-5
EINECS: 205-563-8
UN: 1206
ADR: 3,II
Merck Index: 14,4659
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ff97126664c92362bf0aa2645719548c99f0533&vid=6722523a326fdf1ebabd08a2b8656dd57300787f&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Residual water 0.03%
Residue on Evaporation 3ppm
n-Heptane 96% (info)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR