Kalsiumoksalaatti-H2O ≥99%

Toimittaja: Thermo Fisher Scientific
Varoitus

32430.22 32430.36
32430.22EA 112 EUR
32430.22 32430.36
Kalsiumoksalaatti-H2O ≥99%
Kalsiumoksalaatti-H2O

Purity: 99% CAS No.: 5794-28-5 Molecular Formula: CaC2O4.H2O Formula Weight: 146.11 (128.10anhy) Physical Form: Powder/lump Density: 2.12 Melting Point: 200° Merck Reference: 14,1685 UN No.: 3288 MDL No.: MFCD00150039

Kaava: C₂CaO₄·1H₂O
Sulamispiste: 200 °C
Tiheys: 2,12
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00150039
CAS-numero: 5794-28-5
EINECS: 209-260-1
UN: 3288
ADR: 6.1,III
Merck Index: 14,1685

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR