Dikaliumperoksodisulfaatti ≥99%, BAKER ANALYZED®, J.T.Baker®

Toimittaja: Avantor
Vaara

3238-08EA 0 EUR
3238-08
Dikaliumperoksodisulfaatti ≥99%, BAKER ANALYZED®, J.T.Baker®
Dikaliumperoksodisulfaatti

White powder. Lot analysis on label.

Kaava: K₂S₂O₈
Sulamispiste: 100 °C
Tiheys: 2,477 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011386
CAS-numero: 7727-21-1
EINECS: 231-781-8
UN: 1492
ADR: 5.1,III
Merck Index: 13,07741

Order Now

Testitulosten erittely

For Laboratory,Research,or Manufacturing Use = 99.0 %
ACS - Assay (K₂S₂O₈) (by gravimetry) ≥99.0 %
Insoluble Matter ≤0.005 %
Chloride (Cl) ≤0.001 %
Trace Impurities - Iron (Fe) ≤0.001 %
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Trace Impurities - Manganese (Mn) ≤2.000 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR