Polypropylene centrifuge tubes, PerformR®
Toimittaja: LABCON
3136-345-008-9 3137-345-008-9 3146-335-018-9 3146-870-008-9 3147-335-018-9 3147-870-008-9 3186-345-008-9 3187-345-008-9 3196-335-018-9 3196-870-008-9 3197-335-018-9 3197-870-008-9
3136-345-008-9EA 299 EUR
3136-345-008-9 3137-345-008-9 3146-335-018-9 3146-870-008-9 3147-335-018-9 3147-870-008-9 3186-345-008-9 3187-345-008-9 3196-335-018-9 3196-870-008-9 3197-335-018-9 3197-870-008-9
Polypropylene centrifuge tubes, PerformR®
Putket Sentrifugiputket
PerformR® centrifuge tubes combine reliable performance in major centrifuges with spin rates higher than many popular brands. The high-quality, lab certified, non-cytotoxic medical grade resin means users don't have to compromise their work with low-quality tubes. 
  • ATP-, RNase/Dnase-, and endotoxin-free (non-pyrogenic)
  • Black printed graduations and white writing spot
  • Validated Sterilisation (ISO 11137) for use in Research and Medical labs
  • Polypropylene USP Class VI resin
  • Freezable to –80 °C, and autoclavable
They are designed for everydayMore Product Information