Natriumhydroksidi 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM Reag. Ph. Eur. volumetric solution

Toimittaja: VWR Chemicals
Varoitus

Synonyms: Caustic soda

31770.294EA 31.5 EUR
31770.294 31770.363 31770.374 31770.408 31770.443
Natriumhydroksidi 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM Reag. Ph. Eur. volumetric solution
Natriumhydroksidi
Kaava: NaOH
MW: 40 g/mol
Kiehumispiste: 1390
Sulamispiste: 323
Tiheys: 1.003 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00003548
CAS-numero: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8 III
Merck Index: 14,08627

Order Now

Testitulosten erittely

Titer (20°C; real value 0.2 % accuracy) 0.0998 to 0.1002 mol/l
Conforms to BDH 19145 Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR