Tina(II)klorididihydraatii ≥98%, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Fluka™

Toimittaja: Honeywell Chemicals
Vaara

31669-100G 31669-1KG 31669-500G 31669-6X1KG 31669-25KG
31669-100GEA 60.3 EUR
31669-100G 31669-1KG 31669-500G 31669-6X1KG HONC31669-25KG
Tina(II)klorididihydraatii ≥98%, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Fluka™
Tina(II)klorididihydraatii
Kaava: SnCl₂·2H₂O
Kiehumispiste: Decomposes on heating
Sulamispiste: 37-38 °C
Tiheys: 2,71 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00149863
CAS-numero: 10025-69-1
UN: 3260
ADR: 8,III
Merck Index: 13,08861

Order Now

Testitulosten erittely

Assay 98.0 - 103.0 %
Solubility in HCl complying
Ammonium (NH₄) Max. 0.002 %
Arsenic (As) Max. 0.0001 %
Calcium (Ca) Max. 0.005 %
Copper (Cu) Max. 0.001 %
Iron (Fe) Max. 0.002 %
Potassium (K) Max. 0.005 %
Magnesium (Mg) Max. 0.005 %
Manganese (Mn) Max. 0.0005 %
Sodium (Na) Max. 0.005 %
Nickel (Ni) Max. 0.0005 %
Lead (Pb) Max. 0.005 %
Other heavy metals (as Pb) Max. 0.01 %
Sulfate (SO₄) Max. 0.002 %
Substances not precipitated by H₂S (SO₄) Max. 0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR