Sinkkikloridi ≥98%, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Fluka™

Toimittaja: Honeywell Chemicals
Vaara

31650-1KG 31650-6X250G 31650-25KG
31650-1KGEA 148 EUR
31650-1KG 31650-6X250G 31650-25KG
Sinkkikloridi ≥98%, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Fluka™
Sinkkikloridi
Kaava: ZnCl₂
Sulamispiste: 287…304 °C (1,013 hPa)
Tiheys: 2,93 g/cm³ (22 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011295
CAS-numero: 7646-85-7
EINECS: 231-592-0
UN: 2331
ADR: 8,III
Merck Index: 13,10185

Order Now

Testitulosten erittely

Assay 98.0 - 100.5 %
Insoluble in HCl Max. 0.005 %
pH (10 %, 20°C) 4.6 - 5.5
Ammonium (NH₄) Max. 50 ppm
Calcium (Ca) Max. 0.001 %
Cadmium (Cd) Max. 0.0005 %
Copper (Cu) Max. 0.001 %
Iron (Fe) Max. 0.0005 %
Potassium (K) Max. 0.001 %
Magnesium (Mg) Max. 0.001 %
Sodium (Na) Max. 0.001 %
Lead (Pb) Max. 0.001 %
Al, Ca, Fe, Mg and heavy metals complying
Oxide chloride (as ZnO) complying
Nitrate (NO₃) Max. 0.003 %
Sulfate (SO₄) Max. 20 ppm
Total N Max. 0.002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR