Ceramic tubes, both ends open
Toimittaja: Thermo Fisher Scientific
31606.EH 31606.EO 31609.EH
31606.EHEA 658 EUR
31606.EH 31606.EO 31609.EH
Ceramic tubes, both ends open
Letku Siirtoputki
Pure Al₂O₃ Aluminium oxide AL24.
Slightly porous, good resistance to thermal shock, extremely good creep strength.